![Quetiapine fumarate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/396/110/a20dacf5-c40d-432c-bb9c-717667e038b7/640/a20dacf5-c40d-432c-bb9c-717667e038b7.png)
Quetiapine fumarate
![Quetiapine fumarate European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/396/110/a20dacf5-c40d-432c-bb9c-717667e038b7/640/a20dacf5-c40d-432c-bb9c-717667e038b7.png)
Synonym(s):
Quetiapine hemifumarate salt, 2-[2-(4-Dibenzo[b,f][1,4]thiazepin-11-yl-1-piperazinyl)ethoxy]ethanol hemifumarate, Quetiapine fumarate
Empirical Formula (Hill Notation):
C21H25N3O2S · 0.5C4H4O4
CAS Number:
Molecular Weight:
441.54
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing