![Pyrazinamide Related Compound B United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/405/240/18822993-e670-4b96-ad0e-33212e9e28dd/640/18822993-e670-4b96-ad0e-33212e9e28dd.png)
Pyrazinamide Related Compound B
![Pyrazinamide Related Compound B United States Pharmacopeia (USP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/405/240/18822993-e670-4b96-ad0e-33212e9e28dd/640/18822993-e670-4b96-ad0e-33212e9e28dd.png)
Synonym(s):
Pyrazinecarbonitrile
Empirical Formula (Hill Notation):
C5H3N3
CAS Number:
Molecular Weight:
105.10
Products
Boiling Point (°C)
Formula Weight
Manufacturer
Format
Application
Markush Class
Markush Group
API Family
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing