
Oxaliplatin impurity C

Synonym(s):
(OC-6-33)-[(1R,2R)-Cyclohexane-1,2-diamine-κN,κN′][ethanedioato(2-)-κO1,κO2]dihydroxyplatinum
Empirical Formula (Hill Notation):
C8H16N2O6Pt
CAS Number:
Molecular Weight:
431.30
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing