
Olanzapine impurity A

Synonym(s):
5-Methyl-2-[(2-nitrophenyl)amino]-3-thiophenecarbonitrile
Empirical Formula (Hill Notation):
C12H9N3O2S
CAS Number:
Molecular Weight:
259.28
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing