Narcissoside
Synonym(s):
3,4′,5,7-Tetrahydroxy 3′-methoxyflavone 3-rutinoside, 3-O-Methylquercetin 3-rutinoside, 3-Rutinosylisorhamnetin, Isorhamnetin 3-O-rutinoside, Isorhamnetin 3-rhamnoglucoside, Isorhamnetol 3-rutinoside, Narcissin, Quercetin 3′-methyl ether 3-rutinoside
Empirical Formula (Hill Notation):
C28H32O16
CAS Number:
Molecular Weight:
624.54
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Application
Showing 1-1 of 1