
Modafinil Related Compound B

Synonym(s):
2-[(Diphenylmethyl)sulfonyl]acetamide, 2-(Benzhydrylsulfonyl)acetamide, Modafinil sulfone
Empirical Formula (Hill Notation):
C15H15NO3S
CAS Number:
Molecular Weight:
289.35
Products
Brand
Formula Weight
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing