
Methyl malonyl chloride

Synonym(s):
Methyl chloroformylacetate
Linear Formula:
CH3OCOCH2COCl
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing