![Methyl 3-amino-4-methylthiophene-2-carboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/419/488/b74b9426-f196-46e1-84d3-784009f98a8e/640/b74b9426-f196-46e1-84d3-784009f98a8e.png)
Methyl 3-amino-4-methylthiophene-2-carboxylate
![Methyl 3-amino-4-methylthiophene-2-carboxylate certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/419/488/b74b9426-f196-46e1-84d3-784009f98a8e/640/b74b9426-f196-46e1-84d3-784009f98a8e.png)
Synonym(s):
3-Amino-4-methylthiophenecarboxylic acid methyl ester
Empirical Formula (Hill Notation):
C7H9NO2S
CAS Number:
Molecular Weight:
171.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Product Line
Purity
Quality Segment
Application
Markush Class
Markush Group
Special Grade
Showing 1-2 of 2