
Methenamine

Synonym(s):
Methenamine, Hexamethylenetetramine, 1,3,5,7-Tetraazatricyclo[3.3.1.13,7]decane, Hexamine, Urotropine
Empirical Formula (Hill Notation):
C6H12N4
CAS Number:
Molecular Weight:
140.19
Products
Brand
Formula Weight
Manufacturer
Melting Point (°C)
Physical Form
Agency
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3