
L-Carnitine hydrochloride

Synonym(s):
(−)-β-Hydroxy-γ-(trimethylammonio)butyrate, Vitamin BT
Linear Formula:
(CH3)3N+CH2CH(OH)CH2CO2H · Cl-
CAS Number:
Molecular Weight:
197.66
Products
Product Category
Brand
Biological Source
Color
Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Optical Isomer
Reference Material Type
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing