
L-α-Phosphatidylethanolamine, dioleoyl

Synonym(s):
1,2-Dioleoyl-sn-glycero-3-phosphoethanolamine, (9Z)-9-Octadecenoic acid (1R)-1-[[[(2-aminoethoxy)hydroxyphosphinyl]oxy]methyl]-1,2-ethanediyl ester, 1,2-Di(9Z-octadecenoyl)-sn-glycero-3-phosphoethanolamine, 3-sn-Phosphatidylethanolamine, 1,2-dideoyl, L-β,γ-Dioleoyl-α-cephalin, DOPE, Dioleoyl phosphatidylethanolamine, PE(18:1(9Z)/18:1(9Z))
Empirical Formula (Hill Notation):
C41H78NO8P
CAS Number:
Molecular Weight:
744.03
Products
Product Category
Brand
Biological Source
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing