
Isoprothiolane

Synonym(s):
Diisopropyl 2-(1,3-dithiolan-2-ylidene)malonate
Empirical Formula (Hill Notation):
C12H18O4S2
CAS Number:
Molecular Weight:
290.40
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2