HT-2 Toxin
Synonym(s):
15-Acetoxy-3α,4β-dihydroxy-8α-(3-methylbutyryloxy)-12,13-epoxytrichothec-9-ene
Empirical Formula (Hill Notation):
C22H32O8
CAS Number:
Molecular Weight:
424.48
Products
Product Category
Brand
Formula Weight
Purity
Quality Segment
Application
Showing 1-1 of 1