
Histidine hydrochloride monohydrate

Synonym(s):
L-Histidine monohydrochloride monohydrate, L-α-Amino-β-(4-imidazolyl)propionic acid monohydrochloride
Empirical Formula (Hill Notation):
C6H9N3O2 · HCl · H2O
CAS Number:
Molecular Weight:
209.63
Products
Formula Weight
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing