Skip to Content
Merck
Heptadecane analytical standard

Heptadecane

Heptadecane analytical standard
Linear Formula:
CH3(CH2)15CH3
CAS Number:
Molecular Weight:
240.47
EC Number:

Products

Product Category
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing