
Glycolaldehyde dimer

Synonym(s):
1,4-Dioxane-2,5-diol, 2,5-Dihydroxy-1,4-dioxane, Hydroxyacetaldehyde dimer
Empirical Formula (Hill Notation):
C4H8O4
CAS Number:
Molecular Weight:
120.10
Products
Product Category
Brand
Formula Weight
Physical Form
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing