Skip to Content
Merck
Glycerol formal 47-67% 5-hydroxy-1,3-dioxane basis (GC), 33-53% 4-hydroxymethyl-1,3-dioxolane basis (GC)

Glycerol formal

Glycerol formal 47-67% 5-hydroxy-1,3-dioxane basis (GC), 33-53% 4-hydroxymethyl-1,3-dioxolane basis (GC)
Empirical Formula (Hill Notation):
C4H8O3
CAS Number:
Molecular Weight:
104.10

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Chemical Composition
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade