
Germanium(IV) chloride

Synonym(s):
Germanium tetrachloride, Germanium(IV) chloride
Linear Formula:
GeCl4
CAS Number:
Molecular Weight:
214.45
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Showing 1-2 of 2
Compare
Product Number
Description
Pricing