
Gallium(III) bromide

Synonym(s):
Gallium tribromide
Linear Formula:
GaBr3
Products
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Reagent Type
Core
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing