
Gabapentin Related Compound E

Synonym(s):
1-Carboxycyclohexaneacetic acid
Empirical Formula (Hill Notation):
C9H14O4
CAS Number:
Molecular Weight:
186.21
Products
Brand
Formula Weight
Manufacturer
Physical Form
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2