![Fosinopril sodium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/329/105/7cebb31a-2807-4570-872e-73e4411a5db1/640/7cebb31a-2807-4570-872e-73e4411a5db1.png)
Fosinopril sodium
![Fosinopril sodium British Pharmacopoeia (BP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/329/105/7cebb31a-2807-4570-872e-73e4411a5db1/640/7cebb31a-2807-4570-872e-73e4411a5db1.png)
Synonym(s):
(2S,4S)-4-Cyclohexyl-1-(2-{[2-methyl-1-(propanoyloxy)propoxy](4-phenylbutyl)phosphoryl}acetyl)pyrrolidine-2-carboxylic acid, Acecor, Eosinopril, Fosinopril, Monopril, SQ 28555, Secorvas, Staril
Empirical Formula (Hill Notation):
C30H45NO7P · Na
CAS Number:
Molecular Weight:
585.64
Products
Product Category
Brand
Color
Formula Weight
Manufacturer
Physical Form
Purity
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-5 of 5
Compare
Product Number
Description
Pricing