
Fmoc-Gly-OH

Synonym(s):
Fmoc-glycine, Fmoc-Gly-OH
Empirical Formula (Hill Notation):
C17H15NO4
CAS Number:
Molecular Weight:
297.31
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Product Line
Purity
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Amino Acid
Reaction Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing