![Fluocinolone acetonide for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/794/596/ad75dfb7-4a19-4217-9294-5ff18e5e68a8/640/ad75dfb7-4a19-4217-9294-5ff18e5e68a8.png)
Fluocinolone acetonide for peak identification
![Fluocinolone acetonide for peak identification European Pharmacopoeia (EP) Reference Standard](/deepweb/assets/sigmaaldrich/product/structures/794/596/ad75dfb7-4a19-4217-9294-5ff18e5e68a8/640/ad75dfb7-4a19-4217-9294-5ff18e5e68a8.png)
Synonym(s):
Fluocinolone acetonide, 6α,9α-Difluoro-11β,16α,17α,21-tetrahydroxy-1,4-pregnadiene-3,20-dione, 6α,9α-Difluoro-16α-hydroxyprednisolone 16,17-acetonide, 6α-Fluorotriamcinolone acetonide, Sinalar, Synandone
Empirical Formula (Hill Notation):
C24H30F2O6
CAS Number:
Molecular Weight:
452.49
Products
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing