
Favipiravir

Synonym(s):
6-Fluoro-3-hydroxy-2-pyrazinecarboxamide, 6-fluoro-3,4-dihydro-3-oxo-2-pyrazinecarboxamide, Avigan, T-705, T705
Empirical Formula (Hill Notation):
C5H4FN3O2
CAS Number:
Molecular Weight:
157.10
Products
Brand
Color
Formula Weight
Manufacturer
Physical Form
Product Line
Activity Spectrum
Quality Segment
Application
Mode of Action
Reference Material Type
Special Grade
Showing 1-2 of 2