
Emamectin benzoate

Synonym(s):
(4′’R)-4″-Deoxy-4″-(methylamino)avermectin B1 benzoate
CAS Number:
Products
Product Category
Product Line
Format
Quality Segment
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing