
Dodecyltrimethylammonium hydrogen sulfate

Linear Formula:
CH3(CH2)11N(CH3)3HSO4
CAS Number:
Molecular Weight:
325.51
Products
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing