![Diquat metabolite Dipyridone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/347/762/2e0302cb-999b-40ce-b868-73f7f9ae9f02/640/2e0302cb-999b-40ce-b868-73f7f9ae9f02.png)
Diquat metabolite Dipyridone
![Diquat metabolite Dipyridone certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/347/762/2e0302cb-999b-40ce-b868-73f7f9ae9f02/640/2e0302cb-999b-40ce-b868-73f7f9ae9f02.png)
Synonym(s):
6,7-Dihydrodipyrido[1,2-a:2′,1′-c]pyrazine-4,9-dione, 9,10-Dihydro-8a,10a-diazaphenanthrene-1,8-dione, Diquat dipyridone
Empirical Formula (Hill Notation):
C12H10N2O2
CAS Number:
Molecular Weight:
214.22
Products
Product Category
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing