
Dextromethorphan Impurity A

Synonym(s):
(+)-3-Methoxymorphinan hydrochloride, (+)-cis-1,3,4,9,10,10a-Hexahydro-6-methoxy-2H-10,4a-iminoethanophenanthrene
Empirical Formula (Hill Notation):
C17H23NO · HCl
CAS Number:
Molecular Weight:
293.83
Products
Brand
Formula Weight
Agency
Quality Segment
Application
Package Size
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing