![Cyhalofop-butyl PESTANAL®, analytical standard, mixture of stereoisomers](/deepweb/assets/sigmaaldrich/product/structures/874/520/a9fcf863-531f-4487-bebe-4b7d63bdb42b/640/a9fcf863-531f-4487-bebe-4b7d63bdb42b.png)
Cyhalofop-butyl
![Cyhalofop-butyl PESTANAL®, analytical standard, mixture of stereoisomers](/deepweb/assets/sigmaaldrich/product/structures/874/520/a9fcf863-531f-4487-bebe-4b7d63bdb42b/640/a9fcf863-531f-4487-bebe-4b7d63bdb42b.png)
Synonym(s):
Butyl (R)-2-[4-(4-cyano-2-fluorophenoxy)phenoxy]propionate
Empirical Formula (Hill Notation):
C20H20FNO4
CAS Number:
Molecular Weight:
357.38
Products
Product Category
Brand
Formula Weight
Manufacturer
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-2 of 2