![Crotonaldehyde, mixture of cis and trans ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)](/deepweb/assets/sigmaaldrich/product/structures/113/216/149687fc-b9ce-4f8b-bd99-79dd7730d219/640/149687fc-b9ce-4f8b-bd99-79dd7730d219.png)
Crotonaldehyde, mixture of cis and trans
![Crotonaldehyde, mixture of cis and trans ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)](/deepweb/assets/sigmaaldrich/product/structures/113/216/149687fc-b9ce-4f8b-bd99-79dd7730d219/640/149687fc-b9ce-4f8b-bd99-79dd7730d219.png)
Synonym(s):
2-Butenal, mixture of cis and trans
Linear Formula:
CH3CH=CHCHO
CAS Number:
Molecular Weight:
70.09
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing