
Citicoline sodium salt hydrate

Empirical Formula (Hill Notation):
C14H25N4NaO11P2 · xH2O
Molecular Weight:
510.31 (anhydrous basis)
EC Number:
Products
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing