
Chloramphenicol palmitate nonpolymorph A

Synonym(s):
Chloramphenicol palmitate
Empirical Formula (Hill Notation):
C27H42Cl2N2O6
CAS Number:
Molecular Weight:
561.54
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing