
Carglumic Acid

Synonym(s):
N-Carbamyl-L-glutamic acid
Empirical Formula (Hill Notation):
C6H10N2O5
CAS Number:
Molecular Weight:
190.15
Products
Brand
Formula Weight
Gene Alias
Manufacturer
Agency
Format
Quality Segment
Application
API Family
Special Grade
Showing 1-2 of 2