
Calcium levulinate

Synonym(s):
Calcium levulinate dihydrate, γ-Ketovaleric acid calcium salt, 4-Oxopentanoic acid calcium salt, Levulinic acid calcium salt dihydrate
Linear Formula:
CH3COCH2CH2COO · 1/2Ca · H2O
CAS Number:
Molecular Weight:
153.16
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing