Butafenacil
Synonym(s):
1-(Allyloxycarbonyl)-1-methylethyl 2-chloro-5-[1,2,3,6-tetrahydro-3-methyl-2,6-dioxo-4-(trifluoromethyl)pyrimidin-1-yl]benzoate
Empirical Formula (Hill Notation):
C20H18ClF3N2O6
CAS Number:
Molecular Weight:
474.81
Products
Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1