Boc-D-Lys(2-Cl-Z)-OH
Synonym(s):
Boc-D-Lys(2-Cl-Z)-OH
Empirical Formula (Hill Notation):
C19H27ClN2O6
CAS Number:
Molecular Weight:
414.88
Products
Product Category
Brand
Formula Weight
Physical Form
Product Line
Quality Segment
Functional Group
Application
Optical Isomer
Amino Acid
Reaction Type
Showing 1-1 of 1