Bezafibrate
Synonym(s):
2-[4-[2-(4-Chlorobenzamido)ethyl]phenoxy]-2-methylpropanoic acid
Empirical Formula (Hill Notation):
C19H20ClNO4
CAS Number:
Molecular Weight:
361.82
Products
Product Category
Brand
Formula Weight
Gene Alias
Manufacturer
Physical Form
Purity
Format
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-3 of 3