![Bacitracin A VETRANAL®, analytical standard, main stereoisomer ~50 %](/deepweb/assets/sigmaaldrich/product/structures/123/034/d2f78dea-060e-44c1-a9c5-e2d7f7c0eef2/640/d2f78dea-060e-44c1-a9c5-e2d7f7c0eef2.png)
Bacitracin A
![Bacitracin A VETRANAL®, analytical standard, main stereoisomer ~50 %](/deepweb/assets/sigmaaldrich/product/structures/123/034/d2f78dea-060e-44c1-a9c5-e2d7f7c0eef2/640/d2f78dea-060e-44c1-a9c5-e2d7f7c0eef2.png)
Empirical Formula (Hill Notation):
C66H103N17O16S
Molecular Weight:
1422.69
EC Number:
Products
Brand
Formula Weight
Product Line
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing