
Artemether Related Compound B

Synonym(s):
α-Dihydroartemisinin methyl ether, α-Artemether, (3R,5aS,6R,8aS,9R,10R,12R,12aR)-Decahydro-10-methoxy-3,6,9-trimethyl-3,12-epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin
Empirical Formula (Hill Notation):
C16H26O5
CAS Number:
Molecular Weight:
298.37
Products
Brand
Formula Weight
Manufacturer
Agency
Format
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-2 of 2