
Amoxicillin Related Compound G

Synonym(s):
(2S,5R,6R)-6-{(R)-2-[(R)-2-Amino-2-(4-hydroxyphenyl)acetamido]-2-(4-hydroxyphenyl)acetamido}-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid, 4-OH-phenylglycylamoxicillin
Empirical Formula (Hill Notation):
C24H26N4O7S
CAS Number:
Molecular Weight:
514.55
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing