
Ammonium nitrate-15N2

Synonym(s):
Ammonium-15N nitrate-15N
Linear Formula:
15NH415NO3
CAS Number:
Molecular Weight:
82.03
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Isotope
Application
Markush Class
Markush Group
Showing 1-4 of 4