Skip to Content
Merck
Allyl disulfide ≥80%, FG

Allyl disulfide

Allyl disulfide ≥80%, FG
Synonym(s):
Diallyl disulfide
Linear Formula:
CH2=CHCH2SSCH2CH=CH2
CAS Number:
Molecular Weight:
146.27
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Functional Group
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing