
all-cis-4,7,10,13,16,19-Docosahexaenoic acid methyl ester

Synonym(s):
Fame 22:6n-3, Methyl DHA, Methyl all-cis-4,7,10,13,16,19-docosahexenoate
Empirical Formula (Hill Notation):
C23H34O2
CAS Number:
Molecular Weight:
342.51
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Format
Quality Segment
Functional Group
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing