
Agnus castus fruit dry extract

Synonym(s):
Casticin, 3′,5-Dihydroxy-3,4′,6,7-tetramethoxyflavone, 3,4′,6,7-Tetra-O-methylquercetagetin, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxy-4H-1-benzopyran-4-one, 5-Hydroxy-2-(3-hydroxy-4-methoxyphenyl)-3,6,7-trimethoxychromen-4-one, Quercetagetin 3,4′,6,7-tetramethyl ether, Vitexicarpin
Empirical Formula (Hill Notation):
C19H18O8
CAS Number:
Molecular Weight:
374.34
Products
Formula Weight
Manufacturer
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing