
Acetaldehyde-d4

Synonym(s):
Tetradeuteroacetaldehyde
Linear Formula:
CD3CDO
CAS Number:
Molecular Weight:
48.08
Products
Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Isotope
Markush Class
Markush Group
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing