
6-Diazo-5-oxo-L-norleucine

Synonym(s):
(S)-2-Amino-6-diazo-5-oxocaproic acid, DON
Empirical Formula (Hill Notation):
C6H9N3O3
CAS Number:
Molecular Weight:
171.15
Products
Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Activity Spectrum
Quality Segment
Mode of Action
Optical Isomer
Amino Acid
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing