
4-Nitrophenyl β-D-cellobioside

Synonym(s):
p-Nitrophenyl β-D-cellobioside
Empirical Formula (Hill Notation):
C18H25NO13
CAS Number:
Molecular Weight:
463.39
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Showing 1-1 of 1
Compare
Product Number
Description
Pricing