
4-Hydroxyantipyrine

Synonym(s):
4-Hydroxy-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one, NSC 174055
Empirical Formula (Hill Notation):
C11H12N2O2
CAS Number:
Molecular Weight:
204.23
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing