![3-Methylglutaconic acid, mixture of E and Z isomers ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/145/578/e80f1959-a2ed-4c09-aa3c-f081021c9e50/640/e80f1959-a2ed-4c09-aa3c-f081021c9e50.png)
3-Methylglutaconic acid, mixture of E and Z isomers
![3-Methylglutaconic acid, mixture of E and Z isomers ≥98.0% (HPLC)](/deepweb/assets/sigmaaldrich/product/structures/145/578/e80f1959-a2ed-4c09-aa3c-f081021c9e50/640/e80f1959-a2ed-4c09-aa3c-f081021c9e50.png)
Synonym(s):
3-Methyl-2-pentenedioic acid, NSC 249232
Empirical Formula (Hill Notation):
C6H8O4
CAS Number:
Molecular Weight:
144.13
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing