Skip to Content
Merck
3-Ethoxymethacrolein 96%

3-Ethoxymethacrolein

3-Ethoxymethacrolein 96%
Synonym(s):
3-Ethoxy-2-methylpropenal
Linear Formula:
C2H5OCH=C(CH3)CHO
CAS Number:
Molecular Weight:
114.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing